Anti-EPAC2 Rabbit Polyclonal Antibody
Catalog # PRSI6633
Supplier: ProSci Inc.
Specifications
- Antigen symbol:EPAC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:EPAC2 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human EPAC2.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
EPAC2 Antibody: EPAC2, also known as Rap guanine nuclear exchange factor 4 and cAMPGEF-II, is belongs to a family of cyclic adenosine monophosphate (cAMP) binding proteins with guanine nucleotide exchange factor. Like the related protein EPAC1, EPAC2 signaling plays a role in numerous cellular processes such as integrin-mediated cell adhesion, muscle contraction, learning and memory, cell proliferation, and inflammation. Recent evidence suggests that EPAC2 induces synapse remodeling and depression, with mutations in its gene being found in patients with autism.
Type:
Antigen: EPAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat