Specifications
- Antigen name:Rap guanine nucleotide exchange Factor (GEF) 3
- Antigen symbol:EPAC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:EPAC|EPAC1|HSU79275|bcm910|CAMP-GEFI|MGC21410
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:EPAC1 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human EPAC1.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
EPAC1 Antibody: EPAC1, also known as Rap guanine nuclear exchange factor 3 and cAMPGEF-I, is widely expressed but most prominently in brain, heart, kidney, pancreas, spleen, ovary, thyroid and spinal cord. EPAC1 is a cAMP-binding protein with intrinsic guanine nuclear exchange factor activity that couples cAMP production to the activation of Rap, a GTPase belonging to the Ras family. This activation of Rap influences numerous cellular processes such as integrin-mediated cell adhesion, vascular endothelial barrier formation, and cardiac myocyte gap junction formation. Recently, EPAC1 has been suggested to also be involved in the cAMP-dependent regulation of ion channel formation, intracellular Ca++ signalling, ion transporter activity, and exocytosis.
Type:
Antigen: EPAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat