Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-EBI3 Rabbit Polyclonal Antibody
  PRSI4967
undefined
Anti-EBI3 Rabbit Polyclonal Antibody
  PRSI4967

 

  • Antigen name:
    Epstein-Barr virus induced gene 3
  • Antigen symbol:
    EBI3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    IL35 subunit|IL-27B|EBV-induced gene 3 protein|IL27 subunit|Epstein-Barr virus induced gene 3|interleukin-27 subunit beta|cytokine receptor|IL-27 subunit beta|epstein-Barr virus-induced gene 3 protein|IL27B
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    EBI3 antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human EBI3.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

EBI3 Antibody: EBI3 is a subunit in two distinct heterodimeric cytokines: interleukin-27 (IL-27) and IL-35. Like interleukin-23 (IL-23), IL-27 is a recently discovered member of the IL-6/IL-12 family of proinflammatory and immunoregulatory cytokines. It exists as a heterodimer composed of the p40-related protein EBI3 and an IL-12 p35-related protein termed p28. IL-27 is produced after activation by antigen-presenting cells and induces proliferation of naïve but not memory CD4+ T-cells. It acts by binding to its receptor WSX-1 (also known as TCCR) and gp130 which results in the activation of a Jak/STAT signaling cascade, suggesting the IL-27 is involved in the regulation of immune processes. It has been suggested that IL-27 can also be used as a therapeutic agent against cancer as it can also induce tumor-specific anti-tumor activity mediated through CD8+ T-cells, IFN-gamma, and T-bet. IL-35 is composed of EBI3 and the p35 subunit of IL-12 and has been reported to have therapeutic effects against collagen-induced arthritis by expanding the population of regulatory T cells and suppressing Th17 cells. At least two isoform of EBI3 are known to exist.

Type:
Antigen: EBI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse