Filtration capsules, Sartopore® 2 XLM
Supplier: Sartorius
Certificates
About this item
Sartopore® 2 XLM is designed with finer filter membranes (0,1 µm) for the successful retention of Mycoplasma. The build in 0,2 µm prefilter layer is specifically designed to protect the final 0,1 μm membrane layer in cell culture media filtration applications.
- Very high effective filtration area per filter element allows for >50% more performance
- Available in a broad range of sizes and formats to provide linear scale-up from R and D to process scale
PES membrane; support fleece: polyester/PP; core and end caps: PP; O-rings: silicone; housing: PP; filling bell: PC
These filters can be used for sterilizing grade filtration and Mycoplasma removal from plant peptone or yeast supplemented cell culture media, chemically defined cell culture media, cell culture media components such as serum (e. g. Fetal Calf Serum) and all other cell culture media used in biotech manufacturing.
Certifications: Sartopore® 2 XLM are designed, developed and manufactured in accordance with a ISO 9001 certified Quality Management System. Meet or exceed the requirements for WFI quality standards set by the current USP, ASTM F-838, 21 CFR.
Specifications
- Catalog No:
- SARP5445358M8-OO-A
- SARP5448307H3G-SS
- Type:
- Sartopore® 2 XLM MidiCaps size 8
- Sartopore® 2 XLM T-Style MaxiCaps size 3
- Inlet/outlet:
- Single-stepped hose barb (1/2")/Single-stepped hose barb (1/2")
- Tri-Clamp 50 mm (1 1/2")/Tri-Clamp 50 mm (1 1/2")
- Ø×L:
- 77×161 mm
- 110×874 mm
- Max. pressure:
- 5 bar @ 20 °C; 3 @ 50 °C
- 5 bar @ 20 °C; 3 @ 50 °C
Specifications
Frequently Bought Together