Drug and metabolite standards, butylhydroxytoluene, LoGiCal®
Catalog # PROOLGCFOR0679.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:10 mg
- Standards:Reference standard
- CAS No.:128-37-0
- Description:Butylhydroxytoluene
- Formula:C₁₅H₂₄O
- Description:Butylhydroxytoluene