Piperonyl butoxide tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51-03-6
:
Formula:C₁₉H₃₀O₅
MDL Number:MFCD00005842
:338.44 g/mol
Boiling Pt:155 °C (0.3 mmHg)
Density:1.059
Flash Pt:171 °C (340 °F)
Storage Temperature:Ambient
EINECS:200-076-7
UN:3082
ADR:9,III
Specification Test Results
- Formula Weight:338,44
- Purity:90%
- Refractive Index:1,498
- Assay (GC):>88,0%
Specification Test Results
Related Information
Frequently Bought Together