Order Entry
Netherlands
ContactUsLinkComponent
Ceftazidime Related Compounds, Reference Standard
Ceftazidime Related Compounds, Reference Standard
Supplier:  USP
Certificates
Ceftazidime Related Compounds, Reference Standard
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    1000980-60-8
  • Description:
    Ceftazidime Delta-3-Isomer
  • UN No.:
    3077
  • Formula:
    C₂₂H₃₂N₆O₁₂S₂

Specifications

Related Information

Frequently Bought Together