Order Entry
Netherlands
ContactUsLinkComponent
 
Pramipexole Related Compounds, Reference Standard
 :  USP
 
Pramipexole Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    1001648-71-0
  • Synonym(s):
    (6S,7S)-2-Amino-6-(propylamino)-4,5,6,7-tetrahydrobenzothiazol-7-ol
  • Description:
    Pramipexole Related Compound G
  • Formula:
    C₁₀H₁₉Cl₂N₃S

 

Frequently Bought Together