Piperacillin, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- 1541500.
- 1541529.
- Description:
- Piperacillin
- Piperacillin Related Compound E
- Synonym(s):
- 1-ethylpiperazine-2,3-dione
- CAS No.:
- 66258-76-2
- 59702-31-7
- Formula:
- C₂₃H₂₉N₅O₈S
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1541500.PiperacillinSpecifications:
More
Specifications
Cat. No.1541500.DescriptionPiperacillinSynonym(s)CAS No.66258-76-2FormulaC₂₃H₂₉N₅O₈S - 1541529.Piperacillin Related Compound EPiperacillin Related Compound EUSP reference standardVial25 mgLog in for availability1 * 25 mg1 150,00 €Specifications:
More
Specifications
Cat. No.1541529.DescriptionPiperacillin Related Compound ESynonym(s)1-ethylpiperazine-2,3-dioneCAS No.59702-31-7Formula
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



