Rink amide AM resin LL, beads, Novabiochem® 100 - 200 Mesh
:183599-10-2
:MFCD01656036
:MFCD00240214
- Colour:White to yellow to beige
- Form:Beads
- Loading (photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF):0,20 - 0,45 mmol/g
- Swelling volume (in DMF):Lot specific result
- Total swelling volume in CH₂Cl₂:>5,2 ml/g
- Total swelling volume in DMF:>5,2 ml/g
- Identity:Passes test
- Storage:<30 °C
- Polymer matrix:Copoly (styrene-1% DVB), 100 - 200 mesh
Frequently Bought Together