Nimodipine, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1463858.
- 1463870.
- 1463869.
- Description:
- Nimodipine
- Nimodipine Related Compound A
- Nimodipine Related Compound B
- Synonym(s):
- 2-methoxyethyl-1-methylethyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate
- bis(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
- CAS No.:
- 66085-59-4
- 85677-93-6
- 70172-96-2
- Formula:
- C₂₁H₂₆N₂O₇
- C₂₁H₂₄N₂O₇
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1463858.NimodipineSpecifications:
More
Specifications
Cat. No.1463858.DescriptionNimodipineSynonym(s)CAS No.66085-59-4FormulaC₂₁H₂₆N₂O₇ - 1463870.Nimodipine Related Compound ANimodipine Related Compound AUSP reference standardVial50 mgLog in for availability1 * 50 mg1 180,00 €Specifications:
More
Specifications
Cat. No.1463870.DescriptionNimodipine Related Compound ASynonym(s)2-methoxyethyl-1-methylethyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylateCAS No.85677-93-6FormulaC₂₁H₂₄N₂O₇ - 1463869.Nimodipine Related Compound BNimodipine Related Compound BUSP reference standardVial50 mgLog in for availability1 * 50 mg1 170,00 €Specifications:
More
Specifications
Cat. No.1463869.DescriptionNimodipine Related Compound BSynonym(s)bis(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylateCAS No.70172-96-2Formula



