Alkaline detergent, Divoflow 50
Catalog # DIVY7511265
Supplier: Diversey
Specifications
- Description:Alkaline detergent, Divoflow 50
- Pk:1 Can
- pH range:13,0 (1%)
Specifications
About this item
Divoflow 50 is a low foaming alkaline liquid detergent suitable for CIP, bottle washer and other spray applications. It effectively removes organic soils and prevents the formation of tartar.
- Clear pale yellow liquid
- Versatile
- Provides excellent performance with hard water
Divoflow 50 is highly effective at removing a wide range of organic soils and preventing scale build up and can be used in all food and beverage applications, where high causticity and scale inhibition is required. Highly effective detergent for cleaning smoke chambers with low pressure CIP systems. It gives highly cost effective cleaning performance and helps prevent scale build up, improving operational efficiency. This is a single blended product reducing application complexity.