Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standards, oxacillin, Ph. Eur. RS
Organic reference standards, oxacillin, Ph. Eur. RS
Supplier:  EDQM
Certificates
Organic reference standards, oxacillin, Ph. Eur. RS
Supplier:  EDQM

Specifications

  • CAS No.:
    7240-38-2
  • Formula:
    C₁₉H₁₉N₃O₅S
  • Description:
    Oxacillin for peak identification

Specifications

Frequently Bought Together