Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standards, norethisterone, Ph. Eur. RS
Organic reference standards, norethisterone, Ph. Eur. RS
Supplier:  EDQM
Certificates
Organic reference standards, norethisterone, Ph. Eur. RS
Supplier:  EDQM

Specifications

  • CAS No.:
    68-22-4
  • Formula:
    C₂₀H₂₆O₂
  • Description:
    Norethisterone

Specifications

Frequently Bought Together