
2-(4-Thiazolyl)benzimidazole 98+% (by HPLC)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:148-79-8
:
Formula:C₁₀H₇N₃S
MDL Number:MFCD00005587
:201.25 g/mol
Merck Index:14,9289
Melting Pt:304…305 °C
Storage Temperature:Ambient
EINECS:205-725-8
UN:3077
ADR:9,III
:GHS09
Related Information
Frequently Bought Together