Cobalt(II,III) oxide, powder, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 1688-09
Supplier: Avantor
CAS Number:
Specifications
- Pk:100 lb
- Seq:7167
- Environmentally Preferable:
Specifications
About this item
Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide
:
:240.8 g/mol
CAS Number:1308-06-1
Formula:Co₃O₄
MDL Number:MFCD00010939
Melting Pt:895
Merck Index:13,02454