Calcium acetate hemihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0041.0500
Supplier: Avantor
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:167.18 g/mol
CAS Number:62-54-4
Formula:(H₃CCOO)₂Ca·¹/₂H₂O