Organic reference standards, ferulic acid, Ph. Eur.
Catalog # EDQMY0001013
Supplier: EDQM
CAS Number:
Specifications
- Pk:20 mg
- Standards:Reference standard
- CAS No.:537-98-4
- Description:Ferulic acid, Ph. Eur. standard
- Formula:C₁₀H₁₀O₄
- Description:Ferulic acid