Order Entry
Netherlands
ContactUsLinkComponent
Filtration capsules, AcroPak™ 20
undefined
Filtration capsules, AcroPak™ 20
  • Catalog No:
  • 514-4094
  • 514-4097
  • 516-6945
  • 514-4095
  • 514-4096
  • Type:
  • Fluorodyne® II, non sterile
  • Supor®*
  • Supor® EKV, gamma-irradiated
  • Fluorodyne® II, gamma-irradiated
  • Supor®, non sterile
  • Water flow rate:
  • 26 ml/min (0,1 bar)
  • 40 ml/min (0,1 bar)
  • 26 ml/min (0,1 bar)
  • 40 ml/min (0,1 bar)
  • Filtration area:
  • 20 cm²
  • 20 cm²
  • 20 cm²
  • 20 cm²
  • 20 cm²
  • Inlet/outlet:
  • 6,4 - 12,7 mm Ø stepped hose barb with female Luer slip interior and filling bell outlet
  • 6,4 - 12,7 mm Ø stepped hose barb with female Luer slip interior and filling bell outlet
  • 6,4 - 12,7 mm Ø stepped hose barbs with female slip Luer ID in the hose barb
  • 6,4 - 12,7 mm Ø stepped hose barb with female Luer slip interior and filling bell outlet
  • 6,4 - 12,7 mm Ø stepped hose barb with female Luer slip interior and filling bell outlet
  • Max. temperature:
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • Max. pressure:
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
  • 4,1 bar
Frequently Bought Together