Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standards, adapalenes, Ph. Eur.
Organic reference standards, adapalenes, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, adapalenes, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    106685-40-9
  • Formula:
    C₁₈H₁₈O₁
  • Description:
    Adapalene

Specifications

Frequently Bought Together