Order Entry
Netherlands
ContactUsLinkComponent
Anti-C. jejuni Mouse Monoclonal Antibody [clone: 2E-10.]
Anti-C. jejuni Mouse Monoclonal Antibody [clone: 2E-10.]
  ABNOMAB15098
 :  Abnova
Anti-C. jejuni Mouse Monoclonal Antibody [clone: 2E-10.]
  ABNOMAB15098
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    C. jejuni
  • Clonality:
    Monoclonal
  • Clone:
    2E-10.
  • Host:
    Mouse
  • Isotype:
    IgM kappa
  • Reactivity:
    Bacteria
  • Form:
    Liquid
  • Storage buffer:
    In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
  • Storage temperature:
    Store at 4 °C.
  • Immunogen:
    Total sonicate of Campylobacter jejuni.
  • Purification:
    PEG precipitation
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Mouse monoclonal antibody raised against Campylobacter jejuni.

Type: Primary
Antigen: C. jejuni
Clonality: Monoclonal
Clone: 2E-10.
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Bacteria