Anti-HLA-DR Mouse Monoclonal Antibody (APC-C750) [clone: GRB-1]
ABNOMAB15428
: Abnova
- Antibody type:Primary
- Antigen name:MHC class II, DR
- Antigen symbol:HLA-DR
- Clonality:Monoclonal
- Clone:GRB-1
- Conjugation:APC-C750
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light.
- Immunogen:Mononuclear cell leukemia acute undifferentiated.
- Purification:Affinity purification
- Size:100 AS
- Pk:100 Assays
Mouse monoclonal antibody raised against native human HLA-DR.
Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: GRB-1
Conjugation: APC-C750
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human