Anti-CD63 Mouse Monoclonal Antibody (APC) [clone: TEA3/18]
ABNOMAB15359
: Abnova
- Antibody type:Primary
- Antigen name:CD63 molecule
- Antigen symbol:CD63
- Clonality:Monoclonal
- Clone:TEA3/18
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:967
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C.
- Immunogen:Cell preparation of human cytochrome B enriched cells.
- Purification:Affinity purification
- Size:100 AS
- Pk:100 Assays
Mouse monoclonal antibody raised against human CD63.
Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: TEA3/18
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human