Order Entry
Netherlands
ContactUsLinkComponent
Bone punch, Aesculap®
Bone punch, Aesculap®
Catalog # AESCOK741R
Supplier:  Aesculap
Bone punch, Aesculap®
Catalog # AESCOK741R
Supplier:  Aesculap

Specifications

  • Description:
    Bone punch, downwards cutting, 3,3 mm
  • Length:
    140 mm
  • Pk:
    1

Specifications

About this item