Order Entry
Netherlands
ContactUsLinkComponent
Apron with sleeves, MICROCHEM® 3000, model 215
Apron with sleeves, MICROCHEM® 3000, model 215
Catalog # ANSEYE30W-00215-02
Supplier:  Ansell
undefined
Apron with sleeves, MICROCHEM® 3000, model 215
Catalog # ANSEYE30W-00215-02
Supplier:  Ansell
Specifications
  • Clothing Size:
    S
  • Garment application:
    Chemical protection
  • Colour:
    Yellow
  • Seq:
    01
  • Pk:
    30
Specifications
About this item
This apron is made from an external film barrier layer with two non woven layers underneath. The fabric provides a strong liquid and particle barrier.
  • Velcro fastening to neck Tie fastening at the waist Double cuff with knitted inner cuff Welded and taped seams Multi-layer barrier fabric is effective against numerous chemicals Highly visible: Bright yellow for improved worker safety Lightweight yet durable
Typical applications include: Chemicals, oil and petrochemicals, pharmaceutical, food industry (caustic clean downs), sewage purification installations, industrial and tank cleaning, mining
Certifications: Type 3-B/4-B/5-B, EN 14126, EN 1073-2, EN 1149-5