Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Homogenisers
Homogenisers Bench
Accessories for homogeniser, FastPrep®-96, MP Biomedicals
Accessories for homogeniser, FastPrep®-96, MP Biomedicals
Accessories for homogeniser, FastPrep®-96, MP Biomedicals
ICNA119696161
:
MP Biomedicals
Accessories for homogeniser, FastPrep®-96, MP Biomedicals
ICNA119696161
:
MP Biomedicals
Description:
Ratchet nut assembly
Seq:
053
Pk:
1
For:
FastPrep-96 homogenisers