Order Entry
Netherlands
ContactUsLinkComponent
TLC imaging system, MultiDoc-It™ 130
TLC imaging system, MultiDoc-It™ 130
Catalog # 733-2048
undefined
TLC imaging system, MultiDoc-It™ 130
Catalog # 733-2048
Specifications
  • Description:
    TLC imaging system, MultiDoc-It™ 130
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
For imaging in thin layer chromatography (TLC) applications, with mega pixel resolution digital colour camera for capturing gels, plates and membranes.
  • Sturdy metal construction with large access door
  • Hood enclosure provides a darkroom environment and protects the Digi camera
  • Exclusive capture software enables image acquisition, enhancement and report capabilities
  • Built-in white light; safety switch turns UV off when the door is open
  • W×D×H: 356×323×483 mm
Delivery: MultiDoc-It™ TLC model includes two UVGL-25 UV lamps for placement on two sides of the darkroom. The system is run from a suitable laptop or PC which is not supplied. Please contact us for details on accessories, including printers, converter places, image analysis software.