Order Entry
Netherlands
ContactUsLinkComponent
Trimethylsulphonium hydroxide 0.25 M in methanol, Supelco®
Trimethylsulphonium hydroxide 0.25 M in methanol, Supelco®
Catalog # 92732-10X1ML
Supplier:  Merck
CAS Number:  
undefined
Trimethylsulphonium hydroxide 0.25 M in methanol, Supelco®
Catalog # 92732-10X1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275
  • Pk Info:
    10× 1 ml

Specifications

Specification Test Results

  • Concentration:
    ~0,25 M in methanol
  • Storage temperature:
    2 - 8 °C

Specification Test Results

About this item

 :
 :94.17 g/mol
 :M-Clarity™ quality level = MQ100
Flash Pt:11 °C
CAS Number:17287-03-5
Formula:C₃H₁₀OS
MDL Number:MFCD00216756