Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-FNTB Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-9548R-CY3
 :  Bioss
Anti-FNTB Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-9548R-CY3
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Farnesyltransferase, CAAX Box, Beta
  • Antigen symbol:
    FNTB
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2342
  • Antigen synonyms:
    EC 2.5.1.58|FNTB|FTase-beta|CAAX farnesyltransferase subunit beta|Protein farnesyltransferase subunit beta|RAS proteins prenyltransferase beta
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Mammalian protein farnesyl transferases are heterodimeric proteins containing two nonidentical Alpha and beta subunits that attach farnesyl residues to a cysteine at the fourth position from the COOH terminus of several proteins, including nuclear lamins and p21Ras proteins. The natural substrates contain the Cys-A-A-Xaa recognition sequence, where the A residues are aliphatic and Xaa represents methionine, serine, glutamine or cysteine. The purified farnesyl transferase is an a-b heterodimer. The beta subunit, which is known as FT beta, CAAX farnesyltransferase subunit beta, or Ras proteins prenyltransferase subunit beta, is a 437 amino acid protein that contains five PFTB repeats and binds the peptide substrate. The Alpha subunit is suspected to participate in formation of a stable complex with the substrate farnesyl pyrophosphate.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PGGT1B
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat