Specifications
- Antibody type:Primary
- Antigen name:Farnesyltransferase, CAAX Box, Alpha
- Antigen symbol:FNTA
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2339
- Antigen synonyms:GGTase-I-alpha|FTase-alpha|Type I protein geranyl-geranyltransferase subunit alpha|Ras proteins prenyltransferase alpha|Protein farnesyltransferase/geranylgeranyltransferase type-1 subunit alpha|CAAX farnesyltransferase subunit alpha
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
FNTA, also known as CAAX farnesyltransferase (FTase), attaches a farnesyl group from farnesyl pyrophosphate to cysteine residues at the fourth position from the C terminus of proteins that end in the so-called CAAX box, where C is cysteine, A is usually but not always an aliphatic amino acid, and X is typically methionine or serine. This type of posttranslational modification provides a mechanism for membrane localization of proteins that lack a transmembrane domain. This enzyme has the remarkable property of farnesylating peptides as short as four residues in length that conform to the CAAX consensus sequence. FNTA is also a specific cytoplasmic interactor of the transforming growth factor-beta and activin type I receptors. It is likely to be a key component of the signaling pathway which involves p21ras, an important substrate for farnesyltransferase.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PGGT1A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat