Anti-VAT1L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-8420R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Vesicle Amine Transport 1 Like
- Antigen symbol:VAT1L
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:VAT 1L|VAT1L_HUMAN|Vesicle amine transport protein 1 homolog (T. californica) like.|Synaptic vesicle membrane protein VAT 1 homolog like|Synaptic vesicle membrane protein VAT-1 homolog-like|Probable oxidoreductase KIAA1576|VAT1L
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
VAT1L
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: VAT1L
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat