Anti-NPFF Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-8150R-A647
: Bioss
- Antibody type:Primary
- Antigen name:Neuropeptide FF-amide peptide precursor
- Antigen symbol:NPFF
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:8620
- Antigen synonyms:NPFF_HUMAN.|Neuropeptide FF amide peptide precursor|NPFF protein|NPFF|FMRFamide related peptides precursor|FMRFAL|FMRFamide-related peptides [Precursor]|FMFRamide related peptide precursor
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
FMRFamide related peptides are morphine modulating peptides that have many physiologic effects, including the modulation of morphine induced analgesia, elevation of arterial blood pressure, and increased somatostatin secretion from the pancreas. Neuropeptide FF potentiates and sensitizes ACCN2 and ACCN3 channels.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NPFF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat