Anti-ICMT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-8019R-A555
: Bioss
- Antibody type:Primary
- Antigen name:IsoprenylCysteine carboxyl methyltransferase
- Antigen symbol:ICMT
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:23463
- Antigen synonyms:PCCMT|Prenylated protein carboxyl methyltransferase|pcCMT|Isoprenylcysteine carboxylmethyltransferase|Prenylcysteine carboxyl methyltransferase|PPMT|Protein-S-isoprenylcysteine O-methyltransferase|ICMT
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Catalyzes the post-translational methylation of isoprenylated C-terminal cysteine residues.Tissue specificity:Ubiquitously expressed. Expressed at higher levels in the cerebellum and putamen than in other brain regions. Abundant expression seen in the Purkinje cells and pontine neurons.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ICMT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat