Anti-RASA3 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-5954R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Ras GTPase-activating Protein 3
- Antigen symbol:RASA3
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:22821
- Antigen synonyms:GAPIII|GAP1IP4BP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:510-560/834
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Inhibitory regulator of the Ras-cyclic AMP pathway. Binds inositol tetrakisphosphate (IP4) with high affinity. Might be a specific IP4 receptor.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RASA3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat