Order Entry
Netherlands
ContactUsLinkComponent
Anti-MAP2K5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-MAP2K5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-5431R-A647
Supplier:  Bioss
Anti-MAP2K5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-5431R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-Activated Protein Kinase Kinase 5
  • Antigen symbol:
    MAP2K5
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5607
  • Antigen synonyms:
    PRKMK5|MEK5|MAPKK5|HsT17454
  • Modification:
    phospho-Ser129
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

MEK5 is a dual specificity protein kinase belonging to thr Ser/Thr protein kinase family, (MAP kinase kinase family). It is activated by phosphorylation on Ser/Thr by MAP kinase kinases and interacts specifically with ERK5, and not with another MAP kinase like P38. This kinase specifically interacts with and activates MAPK7/ERK5. This kinase itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). It is not phosphorylated by RAFA, RAFB or RAFC and it may interact with GTPases such as CDC42. The signal cascade mediated by this kinase is involved in growth factor stimulated cell proliferation and muscle cell differentiation. MEK5 is expressed in many adult tissues and is most abundant in heart and skeletal muscle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAP2K5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human