Anti-COL4A3BP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-5044R-A555
: Bioss
- Antibody type:Primary
- Antigen name:Collagen, type IV, alpha 3 (Goodpasture antigen) binding protein
- Antigen symbol:COL4A3BP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10087
- Antigen synonyms:STARD11|CERT|CERTL|GPBP|MRD34
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:575-624/624
- Purification:Purified by Protein A
- Pk:100 µl
Shelters ceramides and diacylglycerol lipids inside its START domain and mediates the intracellular trafficking of ceramides and diacylglycerol lipids in a non-vesicular manner.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: COL4A3BP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity: