Anti-UCHL1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-3806R-CY7
Leverancier: Bioss
Specificaties
- Antibody type:Primary
- Antigen name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen symbol:UCHL1
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7345
- Antigen synonyms:Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:30-80/223
- Purification:Purified by Protein A
- Pk:100 µl
Specificaties
Over dit artikel
Ubiquitin-protein hydrolase involved both in the processing of ubiquitin precursors and of ubiquitinated proteins. This enzyme is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. Also binds to free monoubiquitin and may prevent its degradation in lysosomes. The homodimer may have ATP-independent ubiquitin ligase activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat