Specificaties
- Antibody type:Primary
- Antigen name:Farnesyl Diphosphate Synthase
- Antigen symbol:FDPS
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2224
- Antigen synonyms:FPPS|FPS
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specificaties
Over dit artikel
FDPS is a 419 amino acid enzyme belonging to the FPP/GGPP synthetase family. Localized to cytoplasm and peroxisome, FDPS expression is regulated by phorbol esters and polyunsaturated fatty acids. FDPS assists in cholesterol biosynthesis, post-translational protein modifications and synthesis of steroid hormones in the isoprenoid pathway.FDPS catalyzes the formation of farnesyl diphosphate (FPP), a precursor for several classes of essential metabolites including sterols, dolichols, carotenoids, and ubiquinones. FDPS is inactivated by interferon-induced RSAD2, which may result in the disruption of lipid rafts at the plasma membrane. Existing as a homodimer, FDPS may have anti-viral effects when inactivated by RSAD2. Reduced activity of FDPS in liver may partly be the cause of Zellweger syndrome and neonatal adrenoleukodystrophy, both of which are known to be peroxisomal deficiency diseases.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FDPS
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat