Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic translation termination Factor 1
- Antigen symbol:ERF
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2077
- Antigen synonyms:ETS domain containing transcription factor ERF|Ets2 repressor factor|PE 2|ETS domain-containing transcription factor ERF|PE-2|PE2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
ABT1 (activator of basal transcription 1) is a nuclear protein that associates with the TATA-binding protein (TBP) and enhances basal transcription activity of class II promoters. ABT1 associates with TBP in HeLa nuclear extracts in vitro. Another protein, designated ERF, is a member of the Ets family of transcription factors. The members of the Ets family are grouped because they share a highly conserved DNA binding domain. These factors are involved in growth factor pathways and regulate both proliferation and differentiation. ERF (Ets-2 repressor factor) is a ubiquitously expressed Ets-domain protein that exhibits strong transcriptional repressor activity, suppresses Ets-induced transformation and is regulated by MAPK phosphorylation. ERF transcription may be regulated by Ets-domain proteins. Additionally, modulation of ERF activity is involved in the transcriptional regulation of genes activated during entry into G1 phase.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ERF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat