Specifications
- Antibody type:Primary
- Antigen name:Lymphocyte Antigen 6 Complex Locus C1
- Antigen symbol:LY6C
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Ly6c2|Ly6c|AA959465|CD59|Ly-6C|AA682074
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The gene encoding the mouse alloantigen, Ly-6C, maps to chromosome 15 and encodes a 131 amino acid protein that belongs to the Ly-6 family of glycosyl-phosphatidylinositol (GPI)-linked proteins. Ly-6 family members share amino acid homology throughout a distinctive cysteine rich protein domain that incorporates O-linked carbohydrates. Murine Ly-6 molecules have unique patterns of tissue expression during hematopoiesis from multipotential stem cells to lineage committed precursor cells, and on specific leukocyte subpopulations in the peripheral lymphoid tissues. Ly-6C is predominantly expressed on murine peripheral CD8 T cells. Ly-6C is involved in endothelial adhesion, the killing of target cells by CTLs, inducing TCR-mediated activation of IL-2 and IFN-?production in CD8 T cells and the homing of CD8 T cells. In addition, Ly-6C may act as a signaling molecule of LFA-1 activation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LY6C
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human