Specifications
- Antibody type:Primary
- Antigen name:Calcium Homeostasis Modulator 1
- Antigen symbol:CALHM1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:FAM26C|Protein FAM26C|CALHM1|Calcium homeostasis modulator protein 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
CALHM1 is a 346 amino acid multi-pass endoplasmic reticulum membrane protein that belongs to the FAM26 family. CALHM1 co-localizes with GRP 78 to the endoplasmic reticulum. Predominantly expressed in adult brain, CALHM1 may be a pore-forming ion channel that controls cytosolic Ca2+ permeability and cytosolic Ca2+ concentration in the cell. It is suggested that CALHM1 regulates amyloid precursor protein proteolysis and aggregated amyloid-beta peptides levels in a Ca2+ dependent manner. CALHM1 homomultimerizes and shares strong sequence similarities with the selectivity filter of the NMDA receptor, which generates a large Ca2+ conductance across the plasma membrane. CALHM1 may be a potential factor involved in the pathogenesis of Alzheimer's disease.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CALHM1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat