Anti-Pyruvate Dehydrogenase Mouse Monoclonal Antibody [clone: C-term mAb]
Catalog # BWRLMB0082
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Pyruvate Dehydrogenase
- Clonality:Monoclonal
- Clone:C-term mAb
- Host:Mouse
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the C-terminus of Human Pyruvate Dehydrogenase.
This antibody detects endogenous levels of Pyruvate Dehydrogenase (C-terminus) and does not cross-react with related proteins
Type: Primary
Antigen: Pyruvate Dehydrogenase
Clonality: Monoclonal
Clone: C-term mAb
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse