Anti-p-SHP-1 Rabbit Polyclonal Antibody [clone: Y536]
BWRLBS4169
- Antibody type:Primary
- Antigen symbol:p-SHP-1
- Clonality:Polyclonal
- Clone:Y536
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human SHP-1 around the phosphorylation site of Tyrosine 536.
p-SHP-1 (Y536) pAb detects endogenous levels of SHP-1 protein only when phosphorylated at Tyr536.
Type: Primary
Antigen: p-SHP-1
Clonality: Polyclonal
Clone: Y536
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat