Anti-p-CD95/FAS Rabbit Polyclonal Antibody [clone: Y291]
Catalog # BWRLBS6333
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-CD95/FAS
- Clonality:Polyclonal
- Clone:Y291
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human CD95/FAS around the phosphorylation site of Tyrosine 291.
Phospho-FAS(Y291) pAb detects endogenous levels of Phospho-FAS protein.
Type: Primary
Antigen: p-CD95/FAS
Clonality: Polyclonal
Clone: Y291
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human