Anti-p-Myc Rabbit Polyclonal Antibody [clone: T58]
BWRLBS4128
- Antibody type:Primary
- Antigen symbol:p-Myc
- Clonality:Polyclonal
- Clone:T58
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human Myc around the phosphorylation site of Threonine 58.
p-Myc (T58) pAb detects endogenous levels of proto-oncogene c-Myc protein only when phosphorylated at Thr58.
Type: Primary
Antigen: p-Myc
Clonality: Polyclonal
Clone: T58
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat