Anti-p-Catenin-beta Rabbit Polyclonal Antibody [clone: S33/S37/T41]
Catalogus # BWRLBS5056
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Catenin- beta
- Clonality:Polyclonal
- Clone:S33/S37/T41
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
A phospho specific peptide corresponding to residues surrounding Serine 33 and 37 and threonine 41of human Catenin-beta.
p-beta-catenin (S33/S37/T41) pAb detects endogenous levels of beta-catenin protein only when phosphorylated at Ser33/S37/T41.
Type: Primary
Antigen: p-Catenin-beta
Clonality: Polyclonal
Clone: S33/S37/T41
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat