Anti-p-ASK1 Rabbit Polyclonal Antibody [clone: S83]
BWRLBS5054
- Antibody type:Primary
- Antigen symbol:p-ASK1
- Clonality:Polyclonal
- Clone:S83
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human ASK1 around the phosphorylation site of Serine 83.
p-ASK1 (S83) pAb detects endogenous levels of ASK1 protein only when phosphorylated at Ser83.
Type: Primary
Antigen: p-ASK1
Clonality: Polyclonal
Clone: S83
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human