Order Entry
Netherlands
ContactUsLinkComponent
Anti-p-PKC alpha Rabbit Polyclonal Antibody [clone: T505]
Anti-p-PKC alpha Rabbit Polyclonal Antibody [clone: T505]
Catalog # BWRLBS4441
Anti-p-PKC alpha Rabbit Polyclonal Antibody [clone: T505]
Catalog # BWRLBS4441

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-PKC alpha
  • Clonality:
    Polyclonal
  • Clone:
    T505
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human PKC alpha around the phosphorylation site of Threonine 505.

p-PKC ? (T505) pAb detects endogenous levels of PKC ? protein only when phosphorylated at Thr505.

Type: Primary
Antigen: p-PKC alpha
Clonality: Polyclonal
Clone: T505
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat