Anti-Topo IIIbeta-1 Rabbit Polyclonal Antibody [clone: Q84]
BWRLBS1908
- Antibody type:Primary
- Antigen symbol:Topo III beta-1
- Clonality:Polyclonal
- Clone:Q84
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 51-100 of Human Topo IIIbeta-1.
Topo III?-1 (Q84) pAb detects endogenous levels of Topo III?-1 protein.
Type: Primary
Antigen: Topo IIIbeta-1
Clonality: Polyclonal
Clone: Q84
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse