Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-Rab 40B Rabbit Polyclonal Antibody [clone: S265]
Catalog # BWRLBS3388
Anti-Rab 40B Rabbit Polyclonal Antibody [clone: S265]
Catalog # BWRLBS3388

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Rab 40B
  • Clonality:
    Polyclonal
  • Clone:
    S265
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 250-300 of Human Rab 40B.

Rab 40B (S265) pAb detects endogenous levels of Rab 40B protein.

Type: Primary
Antigen: Rab 40B
Clonality: Polyclonal
Clone: S265
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat